EMAIL THIS PAGE TO A FRIEND
M4512 Sigma-Aldrich

Metrizamide

Grade II, 95-97%, crystalline

Synonym: 2-[3-Acetamido-5-(N-methylacetamido)-2,4,6-triiodobenzamido]-2-deoxy-D-glucose, Metrizamide, mixture of anomers

  • CAS Number 31112-62-6

  • Empirical Formula (Hill Notation) C18H22I3N3O8

  • Molecular Weight 789.10

  •  Beilstein/REAXYS Number 6758226

  •  EC Number 250-475-5

  •  MDL number MFCD00010577

Purchase

Properties

type   Grade II
assay   95-97%
form   crystalline
mp   222-224 °C (dec.) (lit.)
storage temp.   −20°C
SMILES string   CN(C(C)=O)c1c(I)c(NC(C)=O)c(I)c(C(=O)N[C@H]2C(O)O[C@H](CO)[C@@H](O)[C@@H]2O)c1I
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
LZ6650000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?