• USA Home
  • M4519 - Manganese(II) nitrate tetrahydrate

EMAIL THIS PAGE TO A FRIEND
M4519 Sigma-Aldrich

Manganese(II) nitrate tetrahydrate

Purchase

Properties

InChI Key   ALIMWUQMDCBYFM-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Target organs 
Brain
RIDADR 
UN 2724 5.1 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?