• USA Home
  • M4631 - MTH-DL-Hydroxyproline

EMAIL THIS PAGE TO A FRIEND
M4631 Sigma-Aldrich

MTH-DL-Hydroxyproline

  • CAS Number 104809-12-3

  • Empirical Formula (Hill Notation) C7H10N2O2S

  • Molecular Weight 186.23

  •  MDL number MFCD00056894

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?