EMAIL THIS PAGE TO A FRIEND
M5014 Sigma-Aldrich

6-Methylpterine

Synonym: 2-Amino-6-methyl-4-pteridinol

  • CAS Number 708-75-8

  • Empirical Formula (Hill Notation) C7H7N5O

  • Molecular Weight 177.16

  •  MDL number MFCD00012137

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?