• USA Home
  • M5148 - N,N′-Bis(3-maleimidopropionyl)-2-hydroxy-1,3-propanediamine

EMAIL THIS PAGE TO A FRIEND
M5148

N,N′-Bis(3-maleimidopropionyl)-2-hydroxy-1,3-propanediamine

powder

  • CAS Number 115388-98-2

  • Empirical Formula (Hill Notation) C17H20N4O7

  • Molecular Weight 392.36

  •  MDL number MFCD00057896

Purchase

Properties

form   powder
solubility   H2O: 10 mg/mL
storage temp.   −20°C
SMILES string   OC(CNC(=O)CCN1C(=O)C=CC1=O)CNC(=O)CCN2C(=O)C=CC2=O

Description

Application

A water soluble, sulfhydryl-specific homobifunctional cross-linking reagent. Typically coupled to molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates and incorporates 17-atom linker.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?