• USA Home
  • M5796 - Magnesium thiosulfate hexahydrate

EMAIL THIS PAGE TO A FRIEND
M5796 Sigma-Aldrich

Magnesium thiosulfate hexahydrate

BioXtra

Purchase

Properties

product line   BioXtra
impurities   <0.0005% Phosphorus (P)
  <0.1% Insoluble matter
solubility   H2O: 1 M at 20 °C, clear, colorless
cation traces   Al: <0.0005%
  Ca: <0.05%
  Cu: <0.0005%
  Fe: <0.0005%
  K: <0.005%
  NH4+: <0.05%
  Na: <0.005%
  Pb: <0.001%
  Zn: <0.0005%
SMILES string   O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=S
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
OM4610000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?