EMAIL THIS PAGE TO A FRIEND
M5824 Sigma-Aldrich

MBX-102 acid

≥98% (HPLC), powder

Synonym: (aR)-4-chloro-a-[3-(trifluoromethyl)phenoxy]benzeneacetic acid, MBX-102/JNJ39659100

Purchase

Properties

Related Categories Approved Therapeutics/Drug Candidates, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience,
InChI Key   DDTQLPXXNHLBAB-CYBMUJFWSA-N
assay   ≥98% (HPLC)
form   powder
color   white to tan
solubility   DMSO: ≥20 mg/mL
originator   Johnson & Johnson
storage temp.   2-8°C

Description

Packaging

10, 50 mg in glass bottle

Biochem/physiol Actions

MBX-102 is a selective PPAR modulator (SPPARM) and has been shown to inhibit phosphorylation of PPARγ.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Nuclear Receptors (Non-Steroids)

The receptors for several classes of non-steroidal compounds, including retinoids, thyroid hormones and vitamin D, also act as ligand-dependent transcription factors. In addition, a large number of r...
Keywords: Absorption, Apoptosis, Atomic absorption spectroscopy, Cancer, Cardiovascular, Cell proliferation, Clinical, Detoxification, Diabetes, Diseases, Gene expression, Hormones, Ligands, Lipid Metabolism, Metabolism, Metabolites, Obesity, Transcription, Transduction, Vitamins

Nuclear Receptors (PPARs)

Peroxisome proliferator activated receptors (PPARs) are members of the nuclear hormone receptor superfamily of ligand-activated transcription factors that are related to retinoid, steroid and thyroid...
Keywords: Anti-inflammatory agents, Atomic absorption spectroscopy, Cancer, Cell proliferation, Clinical, Diabetes, Diseases, Gene expression, Hormones, Inflammation, Ligands, Lipid Metabolism, Metabolic Pathways, Metabolism, Obesity, Oxidations, RNA immunoprecipitation, Transcription, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?