• USA Home
  • M6130 - 1-(cis-11-Eicosenoyl)-rac-glycerol

EMAIL THIS PAGE TO A FRIEND
M6130 Sigma

1-(cis-11-Eicosenoyl)-rac-glycerol

≥99%, solid

Synonym: Mono-11-eicosenoin, Monoicosenoin

  • CAS Number 62207-88-9

  • Empirical Formula (Hill Notation) C23H44O4

  • Molecular Weight 384.59

Purchase

Properties

assay   ≥99%
form   solid
color   white
storage temp.   −20°C

Description

Packaging

Sealed ampule.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?