• USA Home
  • M6886 - Morphine 3-β-D-glucuronide

EMAIL THIS PAGE TO A FRIEND
M6886 Sigma-Aldrich

Morphine 3-β-D-glucuronide

Synonym: M3G

Purchase

Properties

drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

Description

Biochem/physiol Actions

Metabolite of morphine that is devoid of antinociceptic activity; hyperglycemic and neuroexcitatory effects appear to be mediated via a non-opioid mechanism.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?