EMAIL THIS PAGE TO A FRIEND
M7632 Sigma-Aldrich

Met-Lys formate salt

  • CAS Number 104809-21-4

  • Empirical Formula (Hill Notation) C11H23N3O3S · CH2O2

  • Molecular Weight 323.41

  •  MDL number MFCD00058376

  •  PubChem Substance ID 329817911

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   SGTFBBAHFWVVKS-OZZZDHQUSA-N
storage temp.   −20°C

Description

Packaging

100 mg in poly bottle

Biochem/physiol Actions

Met-Lys is a dipeptide.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?