EMAIL THIS PAGE TO A FRIEND
M7788 Sigma-Aldrich

6α-Mannobiose

≥85% (HPAE)

Synonym: α-D-Man-(1→6)-D-Man, 6-O-α-D-Mannopyranosyl-D-mannopyranose

  • CAS Number 6614-35-3

  • Empirical Formula (Hill Notation) C12H22O11

  • Molecular Weight 342.30

  •  MDL number MFCD00214333

  •  PubChem Substance ID 24897185

Purchase

Properties

Related Categories Glycan Component Classes, Glycobiology, Molecular Biology, N-Glycans, N-Linked Fragments,
assay   ≥85% (HPAE)
form   powder
storage temp.   −20°C
SMILES string   OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O
InChI   1S/C12H22O11/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12/h1,4-12,14-21H,2-3H2
InChI key   AYRXSINWFIIFAE-UHFFFAOYSA-N

Description

General description

6α-Mannobiose is a disaccharide.

Application

6α-Mannobiose has been used in a study to assess the mode of binding of oligosaccharides to the potent antiviral protein griffithsin. It has also been used in a study to investigate the influence of disaccharide structure on prebiotic selectivity in vitro.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Glycan Standards

The analysis of the glycan portion of glycoproteins is necessary for monitoring changes in post-translational modification that occur in disease states, as well as evaluating the consistency of glyco...
BioFiles 2007, 2.1, 18.
Keywords: Applications, Biofiles, Chromatography, Digestions, Electrophoresis, Gel electrophoresis, Gene expression, Glycosylations, High performance liquid chromatography, Mass spectrometry, N-linked glycosylation, Phase transitions, Post translational modifications, Purification, Separation, Sequencing

N-Linked Glycans

N-linked glycosylation, modification, and degradation are involved in a wide variety of processes in all organisms from archaea to eukaryotes. It is the most common covalent protein modification in e...
Glycobiology Analysis Manual, 2nd Edition
Keywords: Catalysis, Degradations, Diseases, Eliminations, Fucosylation, Glycosylations, Metabolism, N-linked glycosylation, Sequences, Substitutions, Type, transformation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?