• USA Home
  • M7907 - N-Acetyl-[Cys4,10, D-Phe7]-α-Melanocyte Stimulating Hormone Fragment 4-13

EMAIL THIS PAGE TO A FRIEND
M7907 Sigma-Aldrich

N-Acetyl-[Cys4,10, D-Phe7]-α-Melanocyte Stimulating Hormone Fragment 4-13

≥95% (HPLC)

  • CAS Number 91050-39-4

  • Empirical Formula (Hill Notation) C61H86N18O13S2

  • Molecular Weight 1343.58

  •  MDL number MFCD00133557

  •  PubChem Substance ID 24897201

Purchase

Description

Amino Acid Sequence

Ac-Cys-Glu-His-Phe-Arg-Trp-Cys-Lys-Pro-Val-NH2 [Disulfide bridge: 4-10]

Biochem/physiol Actions

Cyclic MSH analog that is resistant to tryptic degradation; potent agonist with prolonged melanotropic activity in frog and lizard skin bioassays.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?