• USA Home
  • M8022 - Merrifield’s peptide resin

EMAIL THIS PAGE TO A FRIEND
M8022 Sigma-Aldrich

Merrifield’s peptide resin

200-400 mesh

Synonym: Chloromethylated divinylbenzene cross-linked polystyrene

Purchase

Properties

Related Categories Biochemicals and Reagents, Peptide Synthesis, Peptides More...
extent of labeling   0.4-0.9 Cl per g
matrix   1% cross-linked polystyrene
storage temp.   2-8°C
InChI   1S/C10H10.C9H9Cl/c1-3-9-7-5-6-8-10(9)4-2;1-2-8-5-3-4-6-9(8)7-10/h3-8H,1-2H2;2-6H,1,7H2
InChI key   YDYHOUQBBDDITA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?