• USA Home
  • M8655 - Mast cell degranulating peptide HR1

EMAIL THIS PAGE TO A FRIEND
M8655 Sigma

Mast cell degranulating peptide HR1

≥97% (HPLC)

  • CAS Number 80533-94-4

  • Empirical Formula (Hill Notation) C71H133N19O15

  • Molecular Weight 1492.93

  •  MDL number MFCD00133526

Purchase

Properties

assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C

Description

Amino Acid Sequence

Ile-Asn-Leu-Lys-Ala-Ile-Ala-Ala-Leu-Val-Lys-Lys-Val-Leu-NH2

Biochem/physiol Actions

Peptide from giant hornet Vespa orientalis that has biological effects similar to mast cell degranulating peptide from bee venom.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?