EMAIL THIS PAGE TO A FRIEND
M8897 Sigma-Aldrich

3α-Mannobiose

~95%

Synonym: α-D-Man-(1→3)-D-Man, 3-O-α-D-Mannopyranosyl-D-mannopyranose

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates M-O, Disaccharide More...
description   Mixed anomers
InChI Key   QIGJYVCQYDKYDW-UHFFFAOYSA-N
assay   ~95%
storage temp.   −20°C

Description

Biochem/physiol Actions

Postulated to be the major antigenic determinant in the cell wall of yeast.
An inhibitor of fimbrial lectins from enterobacteria.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?