• USA Home
  • M8904 - 6-Maleimidohexanoic acid

EMAIL THIS PAGE TO A FRIEND
M8904 Sigma-Aldrich

6-Maleimidohexanoic acid

≥98% (HPLC)

Synonym: 6-Maleimidocaproic acid, N-Maleoyl-6-aminocaproic acid, N-(5-Carboxypentyl)maleimide

  • CAS Number 55750-53-3

  • Empirical Formula (Hill Notation) C10H13NO4

  • Molecular Weight 211.21

  •  Beilstein/REAXYS Number 1532405

  •  MDL number MFCD00043140

  •  PubChem Substance ID 329818073

Purchase

Properties

assay   ≥98% (HPLC)
storage temp.   2-8°C
SMILES string   OC(=O)CCCCCN1C(=O)C=CC1=O
InChI   1S/C10H13NO4/c12-8-5-6-9(13)11(8)7-3-1-2-4-10(14)15/h5-6H,1-4,7H2,(H,14,15)
InChI key   WOJKKJKETHYEAC-UHFFFAOYSA-N

Description

Application

Probe for thiol groups (SH-groups) in membrane proteins.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?