• USA Home
  • M9143 - (−)-Menthyloxyacetic acid

EMAIL THIS PAGE TO A FRIEND
M9143 Sigma-Aldrich

(−)-Menthyloxyacetic acid

~97% (TLC), liquid

Synonym: (−)-Menthyl carboxymethyl ether

  • CAS Number 40248-63-3

  • Empirical Formula (Hill Notation) C12H22O3

  • Molecular Weight 214.30

  •  Beilstein/REAXYS Number 2444474

  •  EC Number 254-857-2

  •  MDL number MFCD00001483

Purchase

Properties

InChI Key   CILPHQCEVYJUDN-OUAUKWLOSA-N
assay   ~97% (TLC)
form   liquid
color   yellow
refractive index   n20/D 1.4672 (lit.)
bp   163-164 °C/10 mmHg (lit.)
mp   52-55 °C (lit.)
density   1.01 g/mL at 20 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

M3000 (−)-Menthyloxyacetic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?