• USA Home
  • M9167 - 12-Methoxydodecanoic acid

EMAIL THIS PAGE TO A FRIEND
M9167 Sigma-Aldrich

12-Methoxydodecanoic acid

≥95% (NMR)

  • CAS Number 92169-28-3

  • Empirical Formula (Hill Notation) C13H26O3

  • Molecular Weight 230.34

  •  MDL number MFCD00214335

Purchase

Properties

assay   ≥95% (NMR)
SMILES string   [Na+].COCCCCCCCCCCCC([O-])=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?