• USA Home
  • M9252 - 3-Methylpentanoic acid

EMAIL THIS PAGE TO A FRIEND
M9252 Sigma

3-Methylpentanoic acid

~95% (GC)

Synonym: (±)-3-Methylvaleric acid

  • CAS Number 105-43-1

  • Linear Formula CH3CH2CH(CH3)CH2CO2H

  • Molecular Weight 116.16

  •  Beilstein Registry Number 1720696

  •  EC Number 203-297-7

  •  MDL number MFCD00002729

  •  PubChem Substance ID 329818089

Purchase

Properties

InChI Key   IGIDLTISMCAULB-UHFFFAOYSA-N
assay   ~95% (GC)
refractive index   n20/D 1.416(lit.)
bp   196-198 °C(lit.)
density   0.93 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
UN 3265 8/PG 2
WGK Germany 
3
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

222453 3-Methylvaleric acid, 97% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?