• USA Home
  • M9385 - (S)-(−)-α-Methoxy-α-trifluoromethylphenylacetic acid

EMAIL THIS PAGE TO A FRIEND
M9385 Sigma-Aldrich

(S)-(−)-α-Methoxy-α-trifluoromethylphenylacetic acid

Synonym: (−)-MTPA, Mosher’s acid

  • CAS Number 17257-71-5

  • Empirical Formula (Hill Notation) C10H9F3O3

  • Molecular Weight 234.17

  •  Beilstein/REAXYS Number 4684048

  •  EC Number 241-292-1

  •  MDL number MFCD00064200

Purchase

Properties

InChI Key   JJYKJUXBWFATTE-VIFPVBQESA-N
bp   90 °C/0.1 mmHg
mp   46-49 °C (lit.)

Description

Other Notes

doi:10.1038/nprot.2007.354

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

318035 (S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?