• USA Home
  • M9398 - Methyl 2,3-O-isopropylidene-α-D-mannopyranoside

EMAIL THIS PAGE TO A FRIEND
M9398 Sigma-Aldrich

Methyl 2,3-O-isopropylidene-α-D-mannopyranoside

~98%

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates M-O, Monosaccharide More...
assay   ~98%
storage temp.   2-8°C
SMILES string   COC1OC(CO)C(O)C2OC(C)(C)OC12
InChI   1S/C10H18O6/c1-10(2)15-7-6(12)5(4-11)14-9(13-3)8(7)16-10/h5-9,11-12H,4H2,1-3H3
InChI key   NBCSDARMWTYPFT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?