EMAIL THIS PAGE TO A FRIEND
N1250 Sigma-Aldrich

2-Naphthol

crystalline

Synonym: β-Naphthol, 2-Hydroxynaphthalene

  • CAS Number 135-19-3

  • Linear Formula C10H7OH

  • Molecular Weight 144.17

  •  Beilstein/REAXYS Number 742134

  •  EC Number 205-182-7

  •  MDL number MFCD00004067

Purchase

Properties

vapor density   4.97 (vs air)
vapor pressure   10 mmHg ( 145.5 °C)
form   crystalline
color   white to pink
bp   285-286 °C (lit.)
mp   120-122 °C (lit.)
SMILES string   Oc1ccc2ccccc2c1
InChI   1S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H
InChI key   JWAZRIHNYRIHIV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
UN 3077 9/PG 3
WGK Germany 
2
RTECS 
QL2975000
Flash Point(F) 
307 °F
Flash Point(C) 
153 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

185507 2-Naphthol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?