• USA Home
  • N1253 - DL-Norvaline hydroxamate

EMAIL THIS PAGE TO A FRIEND
N1253 Sigma-Aldrich

DL-Norvaline hydroxamate

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   YWQDCRINEFTEGQ-UHFFFAOYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?