• USA Home
  • N1266 - 2-O-(o-Nitrophenyl)-α-D-N-acetylneuraminic acid

EMAIL THIS PAGE TO A FRIEND
N1266 Sigma-Aldrich

2-O-(o-Nitrophenyl)-α-D-N-acetylneuraminic acid

≥95%

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Chromogenic, Enzyme Substrates, Enzymes, Inhibitors, and Substrates More...
InChI Key   HWBGFIGDTJBVQD-UHFFFAOYSA-N
assay   ≥95%
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?