• USA Home
  • N1509 - S-(4-Nitrobenzyl)glutathione

EMAIL THIS PAGE TO A FRIEND
N1509 Sigma-Aldrich

S-(4-Nitrobenzyl)glutathione

  • CAS Number 6803-19-6

  • Empirical Formula (Hill Notation) C17H22N4O8S

  • Molecular Weight 442.44

  •  MDL number MFCD00057609

  •  PubChem Substance ID 24897494

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Glutathione and Derivatives, Peptides
InChI Key   OAWORKDPTSAMBZ-UHFFFAOYSA-N
storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?