• USA Home
  • N178 - 5-(Nonyloxy)tryptamine hydrogenoxalate

EMAIL THIS PAGE TO A FRIEND
N178 Sigma-Aldrich

5-(Nonyloxy)tryptamine hydrogenoxalate

solid

Synonym: 5-(Nonyloxy)-1H-indole-3-ethanamine hydrogen maleate

  • CAS Number 157798-12-4

  • Empirical Formula (Hill Notation) C19H30N2O · C2H2O4

  • Molecular Weight 392.49

  •  MDL number MFCD00274061

Purchase

Properties

form   solid
color   off-white
solubility   45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 10 mg/mL
  DMSO: 15 mg/mL
  ethanol: 20 mg/mL
  H2O: insoluble
storage temp.   −20°C

Description

Biochem/physiol Actions

Novel, high affinity 5-HT1Dβ serotonin receptor agonist.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?