• USA Home
  • N1880 - 4-Methyl-2-nitrobenzonitrile

EMAIL THIS PAGE TO A FRIEND
N1880 Sigma-Aldrich

4-Methyl-2-nitrobenzonitrile

Synonym: 2-Nitro-p-tolunitrile

Purchase

Properties

mp   97-99 °C (lit.)
storage temp.   2-8°C
SMILES string   Cc1ccc(C#N)c(c1)[N+]([O-])=O
InChI   1S/C8H6N2O2/c1-6-2-3-7(5-9)8(4-6)10(11)12/h2-4H,1H3
InChI key   QGBSLPHQCUIZKK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

214299 4-Methyl-2-nitrobenzonitrile, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?