• USA Home
  • N2036 - Nordihydroguaiaretic acid

EMAIL THIS PAGE TO A FRIEND
N2036 Sigma-Aldrich

Nordihydroguaiaretic acid

>97%

Synonym: 1,4-Bis(3,4-dihydroxyphenyl)-2,3-dimethylbutane, 4,4′-(2,3-Dimethyltetramethylene)dipyrocatechol, Masoprocol, NDGA

  • CAS Number 500-38-9

  • Linear Formula [-CH(CH3)CH2C6H3-1,2-(OH)2]2

  • Molecular Weight 302.36

  •  Beilstein/REAXYS Number 2056826

  •  EC Number 207-903-0

  •  MDL number MFCD00002206

Purchase

Properties

assay   >97%
mp   184-186 °C (lit.)
SMILES string   CC(Cc1ccc(O)c(O)c1)C(C)Cc2ccc(O)c(O)c2
InChI   1S/C18H22O4/c1-11(7-13-3-5-15(19)17(21)9-13)12(2)8-14-4-6-16(20)18(22)10-14/h3-6,9-12,19-22H,7-8H2,1-2H3
InChI key   HCZKYJDFEPMADG-UHFFFAOYSA-N
Gene Information   human ... ALOX15(246), PTGS1(5742), PTGS2(5743)

Description

Biochem/physiol Actions

Lipoxygenase inhibitor; polyphenol-bearing o-dihydroxy (catechol) structure.

Other Notes

Synthetic

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UX1750000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

74540 Nordihydroguaiaretic acid, ≥97.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?