• USA Home
  • N2377 - 2-Nitrophenyl myristate

EMAIL THIS PAGE TO A FRIEND
N2377 Sigma-Aldrich

2-Nitrophenyl myristate

Synonym: Tetradecanoic acid 2-nitrophenyl ester

Purchase

Properties

storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCCC(=O)Oc1ccccc1N(=O)=O
InChI   1S/C20H31NO4/c1-2-3-4-5-6-7-8-9-10-11-12-17-20(22)25-19-16-14-13-15-18(19)21(23)24/h13-16H,2-12,17H2,1H3
InChI key   UTJSENBBKMWKKV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?