• USA Home
  • N2381 - 4-Nitrophenyl 1-thio-β-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
N2381 Sigma-Aldrich

4-Nitrophenyl 1-thio-β-D-glucopyranoside

  • CAS Number 2788-56-9

  • Empirical Formula (Hill Notation) C12H15NO7S

  • Molecular Weight 317.32

  •  MDL number MFCD00069830

Purchase

Properties

storage temp.   −20°C
SMILES string   OCC1OC(Sc2ccc(cc2)N(=O)=O)C(O)C(O)C1O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?