• USA Home
  • N2627 - 2-Nitrophenyl palmitate

EMAIL THIS PAGE TO A FRIEND
N2627 Sigma-Aldrich

2-Nitrophenyl palmitate

Synonym: Hexadecanoic acid 2-nitrophenyl ester

Purchase

Properties

Related Categories Biochemicals and Reagents, Chromogen, Enzyme Substrate, Fatty Acyls, Lipids More...
InChI Key   GOOWMQLOYNJICV-UHFFFAOYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?