• USA Home
  • N3131 - 4-(4-Nitrobenzyl)pyridine

EMAIL THIS PAGE TO A FRIEND
N3131 Sigma-Aldrich

4-(4-Nitrobenzyl)pyridine

  • CAS Number 1083-48-3

  • Empirical Formula (Hill Notation) C12H10N2O2

  • Molecular Weight 214.22

  •  Beilstein/REAXYS Number 170877

  •  EC Number 214-108-2

  •  MDL number MFCD00006445

Purchase

Properties

mp   69-71 °C (lit.)
SMILES string   [O-][N+](=O)c1ccc(Cc2ccncc2)cc1
InChI   1S/C12H10N2O2/c15-14(16)12-3-1-10(2-4-12)9-11-5-7-13-8-6-11/h1-8H,9H2
InChI key   MNHKUCBXXMFQDM-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
UT6360000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

N14204 4-(4-Nitrobenzyl)pyridine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?