• USA Home
  • N3411 - L-Nω-Nitroarginine-2,4-L-diaminobutyric amide di(trifluoroacetate) salt

EMAIL THIS PAGE TO A FRIEND
N3411 Sigma-Aldrich

L-Nω-Nitroarginine-2,4-L-diaminobutyric amide di(trifluoroacetate) salt

≥95% (HPLC), powder

Synonym: (2S)-2-amino-N-[(1S)-3-amino-1-(aminocarbonyl)propyl]-5-[[amino(nitroamino)methyl]amino]-pentamide di(trifluoroacetate) salt

  • CAS Number 244185-39-5

  • Empirical Formula (Hill Notation) C14H24N8O8F6

  • Molecular Weight 546.38

  •  MDL number MFCD05865237

Purchase

Properties

assay   ≥95% (HPLC)
form   powder
solubility   H2O: soluble 50 mg/mL
storage temp.   2-8°C
SMILES string   OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.NCC[C@H](NC(=O)[C@@H](N)CCCNC(=N)N[N+]([O-])=O)C(N)=O

Description

Biochem/physiol Actions

Potent and highly selective inhibitor of nNOS (neuronal nitric oxide synthase).

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?