• USA Home
  • N3630 - Nicotinamide 1,N6-ethenoadenine dinucleotide phosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
N3630 Sigma-Aldrich

Nicotinamide 1,N6-ethenoadenine dinucleotide phosphate sodium salt

≥90%

Synonym: 1,N6-Etheno-NADP, 1,N6-Etheno-nicotinamide adenine dinucleotide phosphate

Purchase

Properties

assay   ≥90%
storage temp.   −20°C

Description

Other Notes

Analog of β-NADP

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?