EMAIL THIS PAGE TO A FRIEND
N4129 Sigma-Aldrich

Nonadecane

~99%, semisolid

  • CAS Number 629-92-5

  • Linear Formula CH3(CH2)17CH3

  • Molecular Weight 268.52

  •  Beilstein/REAXYS Number 1742892

  •  MDL number MFCD00009012

Purchase

Properties

vapor density   9.27 (vs air)
vapor pressure   1 mmHg ( 133.2 °C)
InChI Key   LQERIDTXQFOHKA-UHFFFAOYSA-N
assay   ~99%
form   semisolid
autoignition temp.   446 °F
color   white
bp   330 °C (lit.)
mp   30-34 °C (lit.)
density   0.786 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
211-116-8
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

N28906 Nonadecane, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?