• USA Home
  • N4134 - 2-Methyl-4-nitropyridine N-oxide

EMAIL THIS PAGE TO A FRIEND
N4134 Sigma-Aldrich

2-Methyl-4-nitropyridine N-oxide

powder

Synonym: 4-Nitro-2-picoline N-oxide

  • CAS Number 5470-66-6

  • Empirical Formula (Hill Notation) C6H6N2O3

  • Molecular Weight 154.12

  •  MDL number MFCD00014625

Purchase

Properties

form   powder
color   yellow to light orange
mp   155-159 °C (lit.)
storage temp.   2-8°C
SMILES string   Cc1cc(cc[n+]1[O-])[N+]([O-])=O
InChI   1S/C6H6N2O3/c1-5-4-6(8(10)11)2-3-7(5)9/h2-4H,1H3
InChI key   FTTIAVRPJGCXAC-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
WGK Germany 
3
RTECS 
UT5710000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

454850 2-Methyl-4-nitropyridine N-oxide, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?