• USA Home
  • N4521 - 3-(2-Naphthyl)-L-alanine

EMAIL THIS PAGE TO A FRIEND
N4521 Sigma-Aldrich

3-(2-Naphthyl)-L-alanine

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?