• USA Home
  • N4889 - 4-Nitrophenyl β-D-cellotrioside

EMAIL THIS PAGE TO A FRIEND
N4889 Sigma-Aldrich

4-Nitrophenyl β-D-cellotrioside

  • CAS Number 106927-48-4

  • Linear Formula (β-D-Glc[1→4]-β-D-Glc-[1→4]-β-D-Glc-1→OC6H4-p-NO

  • Molecular Weight 625.53

  •  MDL number MFCD00133737

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

N5759 4-Nitrophenyl β-D-cellobioside, ≥98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?