• USA Home
  • N5125 - Naphthol AS nonanoate

EMAIL THIS PAGE TO A FRIEND
N5125 Sigma-Aldrich

Naphthol AS nonanoate

Purchase

Properties

storage temp.   −20°C
SMILES string   CCCCCCCCC(=O)Oc1cc2ccccc2cc1C(=O)Nc3ccccc3
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?