• USA Home
  • N5512 - 3-(2-Naphthyl)-L-alanine

EMAIL THIS PAGE TO A FRIEND
N5512 Sigma

3-(2-Naphthyl)-L-alanine

  • CAS Number 58438-03-2

  • Empirical Formula (Hill Notation) C13H13NO2

  • Molecular Weight 215.25

  •  MDL number MFCD00066087

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?