• USA Home
  • N5763 - S-(4-Nitrobenzyl)-6-thioguanosine

EMAIL THIS PAGE TO A FRIEND
N5763 Sigma-Aldrich

S-(4-Nitrobenzyl)-6-thioguanosine

≥98%, powder

Synonym: 2-Amino-6-[(4-nitrobenzyl)thio]-9-β-D-ribofuranosylpurine, NBTG, NBTGR

  • CAS Number 13153-27-0

  • Empirical Formula (Hill Notation) C17H18N6O6S

  • Molecular Weight 434.43

  •  MDL number MFCD00005737

Purchase

Properties

assay   ≥98%
form   powder
color   yellow
mp   203-205 °C (lit.)
storage temp.   2-8°C

Description

Biochem/physiol Actions

Potent adenosine uptake inhibitor

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?