• USA Home
  • N6753 - N-(2-Nitrophenylsulfenyl)-L-tryptophan (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
N6753 Sigma-Aldrich

N-(2-Nitrophenylsulfenyl)-L-tryptophan (dicyclohexylammonium) salt

crystalline

  • CAS Number 7675-55-0

  • Empirical Formula (Hill Notation) C17H15N3O4S · C12H23N

  • Molecular Weight 538.70

  •  MDL number MFCD00038479

  •  PubChem Substance ID 24897789

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   QLCIBFSGUWUCDS-UQKRIMTDSA-N
form   crystalline
color   yellow
storage temp.   −20°C

Description

Biochem/physiol Actions

N-(2-Nitrophenylsulfenyl)-L-tryptophan (dicyclohexylammonium) salt is a stable NPS derivative of L-tryptophan.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?