• USA Home
  • N6881 - Nicotinamide hypoxanthine dinucleotide phosphate disodium salt

EMAIL THIS PAGE TO A FRIEND
N6881 Sigma-Aldrich

Nicotinamide hypoxanthine dinucleotide phosphate disodium salt

Grade I, ~95%

Synonym: Deamino NADP, Deamino-triphosphopyridine nucleotide

  • CAS Number 6739-64-6

  • Linear Formula C21H25N6O18P3Na2

  • Molecular Weight 788.35

Purchase

Properties

type   Grade I
assay   ~95%
storage temp.   −20°C

Description

Other Notes

Analog of β-NADP

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?