EMAIL THIS PAGE TO A FRIEND
N7377 Sigma

D-Norvaline

Sigma Grade

Synonym: (R)-2-Aminopentanoic acid, (R)-2-Aminovaleric acid

  • CAS Number 2013-12-9

  • Empirical Formula (Hill Notation) C5H11NO2

  • Molecular Weight 117.15

  •  Beilstein Registry Number 1721161

  •  EC Number 217-936-2

  •  MDL number MFCD00008097

Purchase

Properties

grade   Sigma Grade
mp   ≥300 °C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?