EMAIL THIS PAGE TO A FRIEND
N7390 Sigma-Aldrich

Nle-Sta-Ala-Sta

≥95% (HPLC)

Synonym: Sta = statine =3S,4S-4-Amino-3-hydroxy-6-methylheptanoic acid

  • CAS Number 115388-99-3

  • Empirical Formula (Hill Notation) C25H48N4O7

  • Molecular Weight 516.67

Purchase

Properties

assay   ≥95% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Pepstatin analog which inhibits renin activity.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?