• USA Home
  • N7508 - 1-Naphthyl α-D-galactopyranoside

EMAIL THIS PAGE TO A FRIEND
N7508 Sigma-Aldrich

1-Naphthyl α-D-galactopyranoside

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?