• USA Home
  • N7509 - 4-Nitrophenyl β-D-melibiose

EMAIL THIS PAGE TO A FRIEND
N7509 Sigma-Aldrich

4-Nitrophenyl β-D-melibiose

  • CAS Number 104872-92-6

  • Empirical Formula (Hill Notation) C18H25NO13

  • Molecular Weight 463.39

  •  PubChem Substance ID 329818671

Purchase

Properties

InChI Key   ISCYUJSLZREARS-UHFFFAOYSA-N
storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?