• USA Home
  • N7643 - 4-Nitrophenyl 6-O-β-D-galactopyranosyl-β-D-galacto­pyran­oside

EMAIL THIS PAGE TO A FRIEND
N7643 Sigma-Aldrich

4-Nitrophenyl 6-O-β-D-galactopyranosyl-β-D-galacto­pyran­oside

  • CAS Number 80321-98-8

  • Linear Formula (β-D-Gal-[1→6]-β-D-Gal-1→OC6H4-p-NO

  • Molecular Weight 463.39

  •  MDL number MFCD00083451

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?