• USA Home
  • N7882 - Ethyl 4-nitrosopiperazine-1-carboxylate

EMAIL THIS PAGE TO A FRIEND
N7882 Sigma-Aldrich

Ethyl 4-nitrosopiperazine-1-carboxylate

liquid

Synonym: 4-Nitrosopiperazine-1-carboxylic acid ethyl ester

  • CAS Number 13256-15-0

  • Empirical Formula (Hill Notation) C7H13N3O3

  • Molecular Weight 187.20

  •  MDL number MFCD00057354

Purchase

Properties

form   liquid
color   yellow
storage temp.   2-8°C
SMILES string   CCOC(=O)N1CCN(CC1)N=O
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TL1470000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?