EMAIL THIS PAGE TO A FRIEND
N8507 Sigma-Aldrich

Ethyl nicotinate

Synonym: Nicotinic acid ethyl ester

  • CAS Number 614-18-6

  • Empirical Formula (Hill Notation) C8H9NO2

  • Molecular Weight 151.16

  •  Beilstein/REAXYS Number 122937

  •  EC Number 210-370-7

  •  MDL number MFCD00006389

Purchase

Properties

InChI Key   XBLVHTDFJBKJLG-UHFFFAOYSA-N
color   light yellow
refractive index   n20/D 1.504 (lit.)
bp   223-224 °C(lit.)
mp   8-10 °C (lit.)
density   1.107 g/mL at 25 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
199.4 °F
Flash Point(C) 
93 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

E40609 Ethyl nicotinate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?